missing translation for 'onlineSavingsMsg'
Learn More

[4-(Morpholinomethyl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10351093
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351093 250 mg 250mg
10127512 1 g 1g
10726735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351093 Supplier Thermo Scientific Maybridge Supplier No. CC29309CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 91271-65-7
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD04974050
InChI Key MWVQMAWLNHACQK-UHFFFAOYSA-N
Synonym 4-morpholinomethyl phenyl methanol, 4-morpholin-4-ylmethyl phenyl methanol, zlchem 1288, n-4-hydroxymethylbenzyl morpholine, 4-morpholin-4-ylmethyl-phenyl-methanol, 4-morpholin-4-yl methyl phenyl methanol, 4-morpholin-4-ylmethyl phenyl methan-1-ol
PubChem CID 2795499
IUPAC Name [4-(morpholin-4-ylmethyl)phenyl]methanol
SMILES C1COCCN1CC2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material [4-(Morpholinomethyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.