missing translation for 'onlineSavingsMsg'
Learn More

4-n-Dodecyloxybenzoic acid, 98%

Product Code. 11475327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475327 25 g 25g
11465327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475327 Supplier Thermo Scientific Alfa Aesar Supplier No. B22689.14
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2312-15-4 | C19H30O3 | 306.45 g/mol

4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in N,N-Dimethylformamide.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2312-15-4
Molecular Formula C19H30O3
Molecular Weight (g/mol) 306.45
MDL Number MFCD00002543
InChI Key ALQLYJHDBAKLBB-UHFFFAOYSA-N
Synonym 4-dodecyloxy benzoic acid, p-dodecyloxybenzoic acid, 4-n-dodecyloxybenzoic acid, 4-dodecyloxybenzoic acid, benzoic acid, 4-dodecyloxy, benzoic acid, p-dodecyloxy, p-dodecyloxy benzoic acid, 4-dodecyloxy benzoicacid, pubchem2685, acmc-1crse
PubChem CID 75330
IUPAC Name 4-dodecoxybenzoic acid
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 132°C to 137°C
Quantity 25 g
Beilstein 2215914
Solubility Information Insoluble in water. Soluble in N,N-Dimethylformamide.
Formula Weight 306.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecyloxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.