missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylresorcinol, 99%

Product Code. p-7020557
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11474244

Brand: Thermo Scientific Alfa Aesar A12552.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136-77-6 | C12H18O2 | 194.27 g/mol

4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.

Solubility
Soluble in water. 0.05 g/100 mL (18°C)

Notes
Incompatible with acid chlorides, acid anhydrides, oxidizing agents.  Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-77-6
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00002284
InChI Key WFJIVOKAWHGMBH-UHFFFAOYSA-N
Synonym hexylresorcinol, 4-hexylresorcinol, antascarin, ascaricid, ascarinol, 4-n-hexylresorcinol, oxana, p-hexylresorcinol, adrover, caprokol
PubChem CID 3610
IUPAC Name 4-hexylbenzene-1,3-diol
SMILES CCCCCCC1=CC=C(O)C=C1O

Specifications

Melting Point 64°C to 67°C
Boiling Point 333°C to 335°C
Flash Point >100°C (230°F)
Quantity 5 g
Beilstein 2048312
Merck Index 14,4712
Solubility Information Soluble in water. 0.05g/100 mL (18°C)
Formula Weight 194.27
Percent Purity 99%
Chemical Name or Material 4-n-Hexylresorcinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.