missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonylphenol, 98+%

Product Code. 11497477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497477 5 g 5g
11487477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497477 Supplier Thermo Scientific Alfa Aesar Supplier No. A15609.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-40-5 | C15H24O | 220.356 g/mol

4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

Solubility
Sparingly Soluble (0.020 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-40-5
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00002396
InChI Key IGFHQQFPSIBGKE-UHFFFAOYSA-N
Synonym 4-n-nonylphenol, p-nonylphenol, phenol, 4-nonyl, para-nonylphenol, p-n-nonylphenol, phenol, nonyl, phenol, p-nonyl, nonylphenol mixed, phenol, nonyl derivs., 4-nonyl phenol
PubChem CID 1752
ChEBI CHEBI:34440
IUPAC Name 4-nonylphenol
SMILES CCCCCCCCCC1=CC=C(C=C1)O

Specifications

Melting Point 43°C to 46°C
Density 0.937
Boiling Point 127°C to 130°C (27 mmHg)
Flash Point 150°C (302°F)
Refractive Index 1.511
Quantity 5 g
UN Number UN2430
Beilstein 2047450
Solubility Information Sparingly Soluble (0.020g/L) at 25°C.
Formula Weight 220.36
Percent Purity ≥98%
Chemical Name or Material 4-n-Nonylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.