missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals

Product Code. 11431190
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431190 1 g 1g
15443447 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431190 Supplier Thermo Scientific Alfa Aesar Supplier No. H26318.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52709-84-9
Molecular Formula C21H25N
Molecular Weight (g/mol) 291.44
MDL Number MFCD00075146
InChI Key CSQPODPWWMOTIY-UHFFFAOYSA-N
Synonym 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
PubChem CID 104289
IUPAC Name 4-(4-octylphenyl)benzonitrile
SMILES CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 20°C to 22°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 291.43
Percent Purity 99%
Chemical Name or Material 4'-n-Octylbiphenyl-4-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.