missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzoic acid, 99%

Product Code. 11440003
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440003 25 g 25g
11430003 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440003 Supplier Thermo Scientific Alfa Aesar Supplier No. A12086.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26311-45-5 | C12H16O2 | 192.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26311-45-5
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00002572
InChI Key CWYNKKGQJYAHQG-UHFFFAOYSA-N
Synonym benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
PubChem CID 33479
IUPAC Name 4-pentylbenzoic acid
SMILES CCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Quantity 25 g
Beilstein 2044798
Formula Weight 192.26
Percent Purity 99%
Chemical Name or Material 4-n-Pentylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.