missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxyaniline, 97%

Product Code. 15491238
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491238 1 g 1g
15401248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491238 Supplier Thermo Scientific Alfa Aesar Supplier No. H55096.03
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4469-80-1 | C9H13NO | 151.21 g/mol

4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.

Solubility
Slightly soluble (1.3 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4469-80-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00043621
InChI Key DWOIGSLSPPLRKO-UHFFFAOYSA-N
Synonym p-propoxyaniline, benzenamine, 4-propoxy, 4-n-propoxyaniline, 4-propoxyphenyl amine hydrochloride, 4-propoxyphenylamine, hydrochloride, 4-propyloxyaniline, 4-propoxybenzenamine, 4-propoxy-phenylamine
PubChem CID 78221
IUPAC Name 4-propoxyaniline
SMILES CCCOC1=CC=C(N)C=C1

Specifications

Color Brown
Flash Point 110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble (1.3g/L at 25°C).
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-n-Propoxyaniline
compliance-icons
Product Identifier
  • 4-n-Propoxyaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4-n-Propoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.