missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15419667
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419667 5 g 5g
15409667 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419667 Supplier Thermo Scientific Alfa Aesar Supplier No. H52884.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134150-01-9
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD00859261
InChI Key WLCGYIWOKVWFLB-UHFFFAOYSA-N
Synonym 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
PubChem CID 4100861
IUPAC Name (4-propylphenyl)boronic acid
SMILES CCCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 106°C to 108°C
Quantity 5 g
Formula Weight 164.01
Percent Purity 98%
Chemical Name or Material 4-n-Propylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.