missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 99%

Product Code. 10659801 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10659801

Brand: Thermo Scientific Acros 131620010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 645-56-7 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-56-7
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 22.0°C
Color Yellow
Density 0.9830g/mL
Boiling Point 232.0°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C9H12O
Linear Formula CH3CH2CH2C6H4OH
Refractive Index 1.5220 to 1.5240
Quantity 1 g
Specific Gravity 0.983
Molecular Weight (g/mol) 136.194
Formula Weight 136.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-n-Propylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-n-Propylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.