missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenylacetylene, 97%

Product Code. 11476047
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476047 5 g 5g
11486047 25 g 25g
11496047 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476047 Supplier Thermo Scientific Alfa Aesar Supplier No. B25065.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 62452-73-7 | C11H12 | 144.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 62452-73-7
Molecular Formula C11H12
Molecular Weight (g/mol) 144.217
MDL Number MFCD00173886
InChI Key UVFFOABHOIMLNB-UHFFFAOYSA-N
Synonym 4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
PubChem CID 2775133
IUPAC Name 1-ethynyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C#C

Specifikationer

Density 0.91
Boiling Point 205°C
Flash Point 67°C (153°F)
Refractive Index 1.534
Quantity 5 g
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 4-n-Propylphenylacetylene

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.