missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroanisole, 99+%

Product Code. 10041540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10041540 250 g Glass bottle 250g
10745021 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041540 Supplier Thermo Scientific Acros Supplier No. 147022500
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-17-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007327
InChI Key BNUHAJGCKIQFGE-UHFFFAOYSA-N
Synonym 4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
PubChem CID 7485
ChEBI CHEBI:1911
IUPAC Name 1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 51.0°C to 53.0°C
Color Brown to Green
Density 1.2300g/mL
Boiling Point 260.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4OCH3
Quantity 250 g
Beilstein 06, 230
Merck Index 15, 6671
Specific Gravity 1.23
Solubility Information Solubility in water: 0.468g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 153.14
Percent Purity 99+%
Physical Form Crystals
Chemical Name or Material 4-Nitroanisole, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitroanisole
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.