missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroanisole, 99+%

Product Code. 10745021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10745021 5 g Glass bottle 5g
10041540 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10745021 Supplier Thermo Scientific Acros Supplier No. 147020050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 100-17-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007327
InChI Key BNUHAJGCKIQFGE-UHFFFAOYSA-N
Synonym 4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
PubChem CID 7485
ChEBI CHEBI:1911
IUPAC Name 1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1)[N+]([O-])=O

Specifica

CAS Min % 99.0
CAS Max % 100.0
Melting Point 51.0°C to 53.0°C
Color Brown to Green
Density 1.2300g/mL
Boiling Point 260.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4OCH3
Quantity 5 g
Beilstein 06, 230
Merck Index 15, 6671
Specific Gravity 1.23
Solubility Information Solubility in water: 0.468g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 153.14
Percent Purity 99+%
Physical Form Crystals
Chemical Name or Material 4-Nitroanisole, 99+%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 4-Nitroanisole
Avvertenza
  • Warning
Categoria di pericolo
  • Germ cell mutagenicity Category 2
Indicazioni di pericolo
  • H341-Suspected of causing genetic defects.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Titolo del prodotto
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.