missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzaldehyde, 99%

Product Code. p-7081612
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11434773

Brand: Thermo Scientific Alfa Aesar A11655.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol

4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.

Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-16-8
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007346
InChI Key BXRFQSNOROATLV-UHFFFAOYSA-N
Synonym p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
PubChem CID 541
ChEBI CHEBI:66926
IUPAC Name 4-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 104°C to 106°C
Quantity 25 g
Beilstein 386796
Sensitivity Air Sensitive
Merck Index 14,6587
Solubility Information Soluble in water,ethanol,benzene,glacial acetic acid. Slightly soluble in ether.
Formula Weight 151.12
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzaldehyde
compliance-icons
Product Identifier
  • 4-Nitrobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Nitrobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.