missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonamide, 97%

Product Code. 11419256
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419256 25 g 25g
11409256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419256 Supplier Thermo Scientific Alfa Aesar Supplier No. A18101.14
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6325-93-5 | C6H6N2O4S | 202.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6325-93-5
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007937
InChI Key QWKKYJLAUWFPDB-UHFFFAOYSA-N
Synonym benzenesulfonamide, 4-nitro, p-nitrobenzenesulfonamide, 4-nitrobenzolesulfamide, p-nitrophenylsulfonamide, 4-nitrobenzenesulphonamide, benzenesulfonamide, p-nitro, 4-nitro-benzenesulfonamide, unii-tuf52o8y8b, tuf52o8y8b, 4-nitrobenzene-1-sulfonamide
PubChem CID 22784
IUPAC Name 4-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 178°C to 181°C
Quantity 25 g
Formula Weight 202.19
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.