missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonic acid hydrate, 98%

Product Code. 10741062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10741062 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10741062 Supplier Thermo Scientific Acros Supplier No. 415820250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 138-42-1 | C6H5NO5S | 203.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 138-42-1
CAS Min % 80.0
CAS Max % 100.0
Melting Point 105.0°C to 112.0°C
Color Beige to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H5NO5S
Linear Formula SO3HC6H4NO2
MDL Number MFCD09037353
Quantity 25 g
Synonym 4-nitrobenzenesulfonic acid, benzenesulfonic acid, 4-nitro, p-nitrobenzenesulfonic acid, 4-nitrobenzenesulfonate, p-nitrophenylsulfonic acid, 4-nitrobenzenesulphonic acid, 4-nitro-benzenesulfonic acid, benzenesulfonic acid, p-nitro, ccris 3132, acmc-1bvgi
InChI Key SPXOTSHWBDUUMT-UHFFFAOYSA-N
SMILES OS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitrobenzene-1-sulfonic acid
Molecular Weight (g/mol) 203.17
PubChem CID 8740
Formula Weight 203.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzenesulfonic acid hydrate, pract., 85%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrobenzenesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.