missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoic acid, 99%

Product Code. p-7022118
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11427786

Brand: Thermo Scientific Alfa Aesar A14738.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol

4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-23-7
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007352
InChI Key OTLNPYWUJOZPPA-UHFFFAOYSA-N
Synonym 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
PubChem CID 6108
ChEBI CHEBI:262350
SMILES C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 239°C to 241°C
Density 1.61
Quantity 250 g
Beilstein 973593
Merck Index 14,6589
Solubility Information Very slightly soluble in cold water.
Formula Weight 167.12
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzoic acid
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.