missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoic acid, 99+%

Product Code. 10376480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10376480 25 g Plastic bottle 25g
10532911 100 g Plastic bottle 100g
10755051 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376480 Supplier Thermo Scientific Acros Supplier No. 128460250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-23-7
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007352
InChI Key OTLNPYWUJOZPPA-UHFFFAOYSA-N
Synonym 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
PubChem CID 6108
ChEBI CHEBI:262350
SMILES C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 239.0°C to 242.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CO2H
Quantity 25 g
Beilstein 09,389
Merck Index 14,6589
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/110mL alcohol- 1g/45mL ether,1g/12mL methanol- 1g/20mL acetone,1g/150mL chloroform,slightly soluble in benzene and carbon disulfide
Formula Weight 167.12
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzoic acid, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.