missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzotrifluoride, 96%, Thermo Scientific™

Product Code. 10696154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10696154 1 g Glass bottle 1g
10720901 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10696154

Brand: Thermo Scientific Acros 199260010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-54-0
MDL Number MFCD00007358
InChI Key XKYLCLMYQDFGKO-UHFFFAOYSA-N
Synonym 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
PubChem CID 9821
IUPAC Name 1-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 37.0°C to 42.0°C
Color White to Yellow
Boiling Point 81.0°C to 83.0°C (10.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C7H4F3NO2
Linear Formula O2NC6H4CF3
Quantity 1 g
Beilstein 05, II, 251
Molecular Weight (g/mol) 191.11
Formula Weight 191.11
Percent Purity 96%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material 4-Nitrobenzotrifluoride, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.