missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzotrifluoride, 98%

Product Code. 11392767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392767 1 g 1g
11302777 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392767 Supplier Thermo Scientific Alfa Aesar Supplier No. L15766.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 402-54-0 | C7H4F3NO2 | 191.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-54-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007358
InChI Key XKYLCLMYQDFGKO-UHFFFAOYSA-N
Synonym 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
PubChem CID 9821
IUPAC Name 1-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 38°C to 42°C
Boiling Point 81°C to 83°C (10 mmHg)
Flash Point 88°C (190°F)
Odor Characteristic
Quantity 1 g
UN Number UN3431
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzotrifluoride
compliance-icons
Product Identifier
  • 4-Nitrobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: p-Nitrobenzotrifluoride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.