missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl alcohol, 99%

Product Code. 11472388
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472388 100 g 100g
11462388 25 g 25g
11482388 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472388 Supplier Thermo Scientific Alfa Aesar Supplier No. A15742.22
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 619-73-8 | C7H7NO3 | 153.14 g/mol

4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

Solubility
Soluble in water (2 mg/ml at 20°C).

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-73-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007376
InChI Key JKTYGPATCNUWKN-UHFFFAOYSA-N
Synonym 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m
PubChem CID 69275
ChEBI CHEBI:41214
IUPAC Name (4-nitrophenyl)methanol
SMILES OCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 92°C to 95°C
Boiling Point 185°C (12 mmHg)
Flash Point 180°C (356°F)
Quantity 100 g
Beilstein 1424026
Solubility Information Soluble in water (2mg/ml at 20°C).
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.