missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl alcohol, 99%

Product Code. 10553291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10553291 25 g Glass bottle 25g
10725401 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10553291 Supplier Thermo Scientific Acros Supplier No. 128520250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 619-73-8 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-73-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007376
InChI Key JKTYGPATCNUWKN-UHFFFAOYSA-N
Synonym 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m
PubChem CID 69275
ChEBI CHEBI:41214
IUPAC Name (4-nitrophenyl)methanol
SMILES OCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 92.0°C to 95.0°C
Color Yellow
Boiling Point 185.0°C (12.0 mmHg)
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula O2NC6H4CH2OH
Quantity 25 g
Beilstein 06, 450
Solubility Information Solubility in water: 2g/L (20°C)
Formula Weight 153.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.