missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl bromide, 97+%

Product Code. 11492268
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492268 25 g 25g
11402278 100 g 100g
11412278 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492268 Supplier Thermo Scientific Alfa Aesar Supplier No. A15236.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-11-8 | C7H6BrNO2 | 216.03 g/mol

4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.

Solubility
It hydrolyzes in water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-11-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007373
InChI Key VOLRSQPSJGXRNJ-UHFFFAOYSA-N
Synonym 4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
PubChem CID 66011
ChEBI CHEBI:62822
IUPAC Name 1-(bromomethyl)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 97°C to 101°C
Quantity 25 g
UN Number UN3261
Beilstein 742796
Solubility Information It hydrolyzes in water. Soluble in alcohol and ether.
Formula Weight 216.04
Percent Purity ≥97%
Chemical Name or Material 4-Nitrobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.