missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloride, 99%

Product Code. p-7022823
Click to view available options
Quantity:
10 g
250 g
Unit Size:
10g
250g
This item is not returnable. View return policy

Product Code. 11422398

Brand: Thermo Scientific Alfa Aesar A15749.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-14-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007374
InChI Key KGCNHWXDPDPSBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
PubChem CID 7482
ChEBI CHEBI:87406
IUPAC Name 1-(chloromethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCl)[N+](=O)[O-]

Specifications

Melting Point 70°C to 74°C
Quantity 10 g
UN Number UN3261
Beilstein 387187
Formula Weight 171.58
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.