missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, ≥99.5%, Honeywell™ Fluka™

Product Code. 15686940 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15686940 25 g Glass bottle 25g
15676940 100 g Glass bottle 100g
15696940 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15686940 Supplier Honeywell-Fluka Supplier No. 7356025G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 116°C
Boiling Point 279°C
Linear Formula O2NC6H4OH
UN Number UN1663
Beilstein 1281877
Solubility Information 14.8g/L at 25°C
Formula Weight 139.11g/mol
Percent Purity ≥99.5%
Packaging Glass bottle
Vapor Pressure 0.6mmHg (120°C)
Density 1.479g/cm3
Color Yellow
pH 4.4
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material 4-Nitrophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.