missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, Extra Pure, SLR, Ph Indicator, Fisher Chemical

Product Code. 10040280 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10040280 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10040280 Supplier Fisher Chemical Supplier No. N/3200/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 100-02-7
Melting Point 112°C
Color Brown
pH 4.4
Boiling Point 279°C
Packaging Glass Jar
Molecular Formula C6H5NO3
MDL Number MFCD00007331
Quantity 100 g
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitrophenol
Molecular Weight (g/mol) 139.11
PubChem CID 980
ChEBI CHEBI:16836
Formula Weight 139.11g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.