missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, 95%, Thermo Scientific Chemicals

Catalog Number 11936011
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
80149-80-0
C12H17NO5Si
283.36
MFCD00042930
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
3086116
(4-nitrophenyl) 2-trimethylsilylethyl carbonate
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
80149-80-0
C12H17NO5Si
283.36
MFCD00042930
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
3086116
(4-nitrophenyl) 2-trimethylsilylethyl carbonate
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 38°C to 39°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 283.36
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'