missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Acetate, TRC

Product Code. 30472843
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30472843 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472843 Supplier Toronto Research Chemicals Supplier No. TRCN4969855G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-03-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
Synonym NSC 2633, p-Acetoxynitrobenzene, p-Nitrobenzene Acetate, p-Nitrophenol Acetate, p-Nitrophenyl Acetate, p-Nitrophenyl Ester Acetic Acid, p-Nitrophenol Acetate
IUPAC Name (4-nitrophenyl) acetate
SMILES CC(=O)Oc1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H7NO4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3
Formula Weight 181.0375
Chemical Name or Material 4-Nitrophenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.