missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl benzoate, 97%

Product Code. 11342867
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11342867

Brand: Thermo Scientific Alfa Aesar L16024.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 959-22-8 | C13H9NO4 | 243.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-22-8
Molecular Formula C13H9NO4
Molecular Weight (g/mol) 243.218
MDL Number MFCD00135494
InChI Key GMKZBFFLCONHDE-UHFFFAOYSA-N
Synonym p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester
PubChem CID 70396
IUPAC Name (4-nitrophenyl) benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 142°C to 144°C
Quantity 25 g
Beilstein 2054709
Formula Weight 243.22
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.