missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Chloroformate >90%, TRC

Product Code. 18063410
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063410 100 g 100g
18081110 25 g 25g
17909629 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18063410 Supplier Toronto Research Chemicals Supplier No. TRCN503310100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7693-46-1
Formula Weight 200.9829
InChI Formula InChI=1S/C7H4ClNO4/c8-7(10)13-6-3-1-5(2-4-6)9(11)12/h1-4H
IUPAC Name (4-nitrophenyl) carbonochloridate
Molecular Formula C7 H4 Cl N O4
Molecular Weight (g/mol) 201.564
SMILES [O-][N+](=O)c1ccc(OC(=O)Cl)cc1
Synonym Carbonochloridic Acid 4-Nitrophenyl Ester, (4-Nitrophenoxy)carbonyl Chloride, (p-Nitrophenoxy)carbonyl Chloride, 4-Nitrophenyl Carbonochloridate, 4-Nitrophenyl Chlorocarbonate, Chloroformic Acid 4-nitrophenyl Ester, Chloroformic Acid p-Nitrophenyl Ester, p-Nitrophenol Chloroformate, p-Nitrophenyl Chlorocarbonate, p-Nitrophenyl Chloroformate
Chemical Name or Material 4-Nitrophenyl Chloroformate

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.