missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitrotoluene, 99%

Product Code. 11457007
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457007 100 g 100g
11467007 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11457007 Supplier Alfa Aesar™ Supplier No. A14398.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Applications
4-Nitrotoluene is used as a basic chemical in the chemical industry for the manufacturing of intermediates.

Solubility
Soluble in water at 20°C 0.26 g/L. Soluble in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-99-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007366
InChI Key ZPTVNYMJQHSSEA-UHFFFAOYSA-N
Synonym 4-nitrotoluene, p-nitrotoluene, 4-methylnitrobenzene, 4-nitrotoluol, benzene, 1-methyl-4-nitro, toluene, p-nitro, p-methylnitrobenzene, para-nitrotoluol, nitrotoluenos, nitrotoluene, p
PubChem CID 7473
ChEBI CHEBI:35227
IUPAC Name 1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 50°C to 54°C
Boiling Point 238°C to 239°C
Flash Point 106°C (222°F)
Odor Almond-like
Quantity 100 g
UN Number UN3446
Beilstein 1906911
Merck Index 14,6650
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material 4-Nitrotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.