missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroveratrole, 98+%

Product Code. 11335169
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335169 250 g 250g
11315169 25 g 25g
11325169 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335169 Supplier Thermo Scientific Alfa Aesar Supplier No. A18264.30
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 709-09-1 | C8H9NO4 | 183.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 709-09-1
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD00007238
InChI Key YFWBUVZWCBFSQN-UHFFFAOYSA-N
Synonym 3,4-dimethoxynitrobenzene, 4-nitroveratrole, benzene, 1,2-dimethoxy-4-nitro, 4-nitrcveratrole, 4-nitro-1,2-dimethoxybenzene, benzene,2-dimethoxy-4-nitro, 1,2-dimethoxy-4-nitrobenzol, 3,4-dimethoxylnitrobenzene, 3,4-dimethoxy-1-nitrobenzene
PubChem CID 69728
IUPAC Name 1,2-dimethoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 95°C to 98°C
Boiling Point 221°C to 223°C (10 mmHg)
Quantity 250 g
Beilstein 1878614
Formula Weight 183.16
Percent Purity ≥98%
Chemical Name or Material 4-Nitroveratrole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.