missing translation for 'onlineSavingsMsg'
Learn More

4-Octylbenzaldehyde, 97%

Product Code. 10764563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10764563 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10764563 Supplier Thermo Scientific Acros Supplier No. 368880050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49763-66-8 | C15H22O | 218.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 49763-66-8
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C15H22O
Linear Formula CHOC6H4CH2(CH2)6CH3
Refractive Index 1.5100 to 1.513
MDL Number MFCD00041872
Quantity 5 g
Synonym benzaldehyde, 4-octyl, n-octylbenzaldehyde, 4-n-octylbenzaldehyde, p-octylbenzaldehyde, paraoctylbenzaldehyde, p-n-octylbenzaldehyde, para-octylbenzaldehyde, p-octylbenzaldehyde, l
InChI Key IHKVZPVHTKOSLW-UHFFFAOYSA-N
SMILES CCCCCCCCC1=CC=C(C=C1)C=O
IUPAC Name 4-octylbenzaldehyde
Molecular Weight (g/mol) 218.34
PubChem CID 2734685
Formula Weight 218.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Octylbenzaldehyde, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.