missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol Dihydrochloride, TRC

Product Code. 30007010
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007010 500 mg 500mg
17904595 1 g 1g
17940665 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007010 Supplier Toronto Research Chemicals Supplier No. TRCC377570500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164726-80-1
Molecular Formula C19 H23 Cl N2 O . 2 Cl H
Molecular Weight (g/mol) 403.77
Synonym 2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol Dihydrochloride, Cetirizine Dihydrochloride Imp. G (EP) as Dihydrochloride, 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazine-ethanol Dihydrochloride, 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-, hydrochloride (1:2), 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-, dihydrochloride (9CI)
IUPAC Name 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol;dihydrochloride
SMILES Cl.Cl.OCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H23ClN2O.2ClH/c20-18-8-6-17(7-9-18)19(16-4-2-1-3-5-16)22-12-10-21(11-13-22)14-15-23;;/h1-9,19,23H,10-15H2;2*1H
Formula Weight 402.1032
Chemical Name or Material 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.