missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)phenol, 97%

Product Code. 15416227
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416227 1 g 1g
15426227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416227 Supplier Thermo Scientific Alfa Aesar Supplier No. H33413.03
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 774-94-7 | C6H5F5OS | 220.16 g/mol

Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-94-7
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788516
InChI Key XHJLGVIUMCBMHL-UHFFFAOYSA-N
Synonym 4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
PubChem CID 2779203
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Quantity 1 g
Formula Weight 220.16
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)phenol
compliance-icons
Product Identifier
  • 4-(Pentafluorothio)phenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Hydroxyphenylsulfur pentafluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.