missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10757465
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10757465 25 g 25g
10566353 1 g 1g
10270163 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10757465 Supplier Thermo Scientific Maybridge Supplier No. BTB09863EB
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26311-45-5
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00002572
InChI Key CWYNKKGQJYAHQG-UHFFFAOYSA-N
Synonym benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
PubChem CID 33479
IUPAC Name 4-pentylbenzoic acid
SMILES CCCCCC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH3(CH2)4C6H4CO2H
Quantity 25 g
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.