missing translation for 'onlineSavingsMsg'
Learn More

4'-Phenoxyacetophenone, 98+%

Product Code. 11469553
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469553 100 g 100g
11459553 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469553 Supplier Thermo Scientific Alfa Aesar Supplier No. A12392.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5031-78-7 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5031-78-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008744
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
Synonym 4'-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one, ethanone, 1-4-phenoxyphenyl, 4-acetyldiphenyl ether, 1-4-phenoxyphenyl-1-ethanone, 1-4-phenoxyphenyl ethanon, pubchem13195
PubChem CID 236783
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 325°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2049295
IUPAC Name 1-(4-phenoxyphenyl)ethan-1-one
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material 4'-Phenoxyacetophenone
compliance-icons
Product Identifier
  • 4'-Phenoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Phenoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.