missing translation for 'onlineSavingsMsg'
Learn More

4'-Phenoxyacetophenone, 98+%

Product Code. 15413057
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413057 Supplier Thermo Scientific Alfa Aesar Supplier No. A12392.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5031-78-7 | C14H12O2 | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5031-78-7
Melting Point 48°C to 52°C
Boiling Point 325°C
Flash Point >110°C
Molecular Formula C14H12O2
MDL Number MFCD00008744
Quantity 5 g
Beilstein 2049295
Synonym 4 '-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
IUPAC Name 1-(4-phenoxyphenyl)ethan-1-one
Molecular Weight (g/mol) 212.25
PubChem CID 236783
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material 4 '-Phenoxyacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.