missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzaldehyde, 98%

Product Code. 11476036
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476036 25 g 25g
11456036 1 g 1g
11466036 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476036 Supplier Thermo Scientific Alfa Aesar Supplier No. B22329.14
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-36-7 | C13H10O2 | 198.22 g/mol

4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-36-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00003383
InChI Key QWLHJVDRPZNVBS-UHFFFAOYSA-N
Synonym p-phenoxybenzaldehyde, 4-formyldiphenyl ether, benzaldehyde, p-phenoxy, benzaldehyde, 4-phenoxy, 4-phenoxy-benzaldehyde, 4-phenyloxy benzaldehyde, diphenyl ether 4-carboxaldehyde, zlchem 548, 4-formyldiphenylether, 4-phenoxy benzaldehyde
PubChem CID 66139
IUPAC Name 4-phenoxybenzaldehyde
SMILES O=CC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 24°C to 25°C
Density 1.132
Boiling Point 184°C to 186°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.611
Quantity 25 g
Beilstein 1947841
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.