missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxybenzenesulfonyl chloride, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS | 1623-92-3 |
| Assay Percent Range | 96% min. diethylsulfonamide derivative (HPLC) |
| Packaging | Glass Bottle |
| Molecular Formula | C12H9ClO3S |
| MDL Number | MFCD00625748 |
| Quantity | 1g |
| Synonym | 4-phenoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-phenoxy, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxyphenylsulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxy benzene sulfonyl chloride, acmc-1bvtb, chloro 4-phenoxyphenyl sulfone, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride |
| InChI Key | QIZPONOMFWAPRR-UHFFFAOYSA-N |
| SMILES | ClS(=O)(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 |
| IUPAC Name | 4-phenoxybenzenesulfonyl chloride |
| Show More |
Safety and Handling
Product Identifier
- 4-Phenoxybenzenesulfonyl chloride, 97%
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?