missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzonitrile, ≥97%, Maybridge

Product Code. 10337504
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337504 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10337504 Supplier Thermo Scientific Maybridge Supplier No. CC53716DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3096-81-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H9NO
MDL Number MFCD00017346
Quantity 1 g
Synonym benzonitrile, 4-phenoxy, 4-phenoxybenzenecarbonitrile, 4-cyanodiphenyl ether, acmc-209hjp, 4-phenoxybenzonitrile, ksc497e6d
InChI Key UYHCIOZMFCLUDP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C#N
IUPAC Name 4-phenoxybenzonitrile
Molecular Weight (g/mol) 195.221
PubChem CID 137821
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.