missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzyl bromide, 97%

Product Code. 15496777
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496777 1 g 1g
15406787 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496777 Supplier Thermo Scientific Alfa Aesar Supplier No. H33895.03
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36881-42-2 | C13H11BrO | 263.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36881-42-2
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD04038693
InChI Key CPIGBCFBFZSCQI-UHFFFAOYSA-N
PubChem CID 1514251
IUPAC Name 1-(bromomethyl)-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr

Specifications

Quantity 1 g
UN Number UN3265
Synonym 1-bromomethyl-4-phenoxybenzene, 4-phenoxybenzyl bromide, 1-bromomethyl-4-phenoxy-benzene, benzene,1-bromomethyl-4-phenoxy, 4-phenyloxybenzylbromide, 4-bromomethyl-1-phenoxybenzene, benzene, 1-bromomethyl-4-phenoxy
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.