missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10182794
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10182794 250 mg 250mg
10092014 1 g 1g
10327684 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182794 Supplier Thermo Scientific Maybridge Supplier No. CC63701CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 31719-76-3
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00623268
InChI Key GRBUVHSYBRTCIB-UHFFFAOYSA-M
Synonym 4-phenoxymethyl benzoic acid, 4-phenoxymethyl benzenecarboxylic acid, 4-phenoxymethyl-benzoic acid, phenoxymethylbenzenecarboxylicacid
PubChem CID 737267
IUPAC Name 4-(phenoxymethyl)benzoic acid
SMILES [O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 228.25
Percent Purity 97%
Chemical Name or Material 4-(Phenoxymethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.