missing translation for 'onlineSavingsMsg'
Learn More

[4-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10347254
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347254 1 g Amber Glass Bottle 1g
10225904 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347254 Supplier Thermo Scientific Maybridge Supplier No. CC63709DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 262862-97-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD09879941
InChI Key MTQHORUPINPVTP-UHFFFAOYSA-N
Synonym 4-phenoxymethyl phenyl methanol, 4-phenoxymethylphenylmethanol, 4-phenoxymethylbenzyl alcohol, 4-phenoxymethylphenyl-methanol, 4-phenoxymethyl phenyl methan-1-ol
PubChem CID 11333367
IUPAC Name [4-(phenoxymethyl)phenyl]methanol
SMILES C1=CC=C(C=C1)OCC2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material [4-(Phenoxymethyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.