missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenylboronic acid, 95+%

Product Code. 10714014 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10714014 25 g Glass bottle 25g
10613763 1 g Glass bottle 1g
10668794 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10714014 Supplier Thermo Scientific Acros Supplier No. 359640250
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51067-38-0 | C12H11BO3 | 214.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51067-38-0
Molecular Formula C12H11BO3
Molecular Weight (g/mol) 214.03
MDL Number MFCD00093312
InChI Key KFXUHRXGLWUOJT-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
PubChem CID 2734377
IUPAC Name (4-phenoxyphenyl)boronic acid
SMILES OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 141°C to 145°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 214.02
Percent Purity 95+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.