missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Product Code. 10646820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Conditionnement:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Packaging unitSize
10646820 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10646820 Fournisseur Thermo Scientific Acros Code fournisseur 431720500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 21473-02-9 | C12H9BrMgO | 273.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spécification

CAS 21473-02-9, 109-99-9
CAS Min % 11.29, 83.07
CAS Max % 16.93, 88.71
Color Yellow
Density 0.9680g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C12H9BrMgO
MDL Number MFCD00672009
Quantity 50 mL
Specific Gravity 0.968
Synonym 4-phenoxyphenylmagnesium bromide, p-phenoxyphenylmagnesium bromide, p-phenoxyphenyl magnesium bromide, 4-phenoxy-phenylmagnesium bromide, 4-phenoxyphenyl magnesium bromide, dtikyzptjrzftl-uhfffaoysa-m, magnesium,bromo 4-phenoxyphenyl, bromo 4-phenoxyphenyl magnesium, 4-phenoxyphenylmagnesium bromide solution, 4-phenoxyphenylmagnesium bromide, 0.5 m in 2-methf, grignard reagent
InChI Key DTIKYZPTJRZFTL-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=C(OC2=CC=CC=C2)C=C1
IUPAC Name magnesium;phenoxybenzene;bromide
Molecular Weight (g/mol) 273.41
PubChem CID 13467796
Formula Weight 273.14
Physical Form Solution
Chemical Name or Material 4-Phenoxyphenylmagnesium bromide
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 4-Phenoxyphenylmagnesium bromide, 0,5M solution in THF
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Mention de danger
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.