missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-buten-2-one, 98+%, Thermo Scientific Chemicals

Product Code. 10616614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616614 50 g Glass bottle 50g
10510184 250 g Glass bottle 250g
10519993 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616614 Supplier Thermo Scientific Acros Supplier No. 385590500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-57-6 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-57-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00008779
InChI Key BWHOZHOGCMHOBV-BQYQJAHWSA-N
Synonym benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton
PubChem CID 637759
ChEBI CHEBI:78399
IUPAC Name (E)-4-phenylbut-3-en-2-one
SMILES CC(=O)\C=C\C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 38.5°C
Color Yellow
Boiling Point 261°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCOCH3
Quantity 50 g
Merck Index 15, 1140
Solubility Information Solubility in water: 1.3g/L (20°C). Other solubilities: sparingly soluble in petroleum ether,freely soluble in alcohol,benzene,chloroform,,ether
Formula Weight 146.19
Percent Purity 98+%
Physical Form Low Melting Solid
Chemical Name or Material 4-Phenyl-3-buten-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenyl-3-buten-2-one
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.