missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%

Product Code. 10275070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275070 25 g Glass bottle 25g
10407320 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275070 Supplier Thermo Scientific Acros Supplier No. 122920250
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 83949-32-0 | C12H15NO2·C7H8O3S | 377.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83949-32-0
Molecular Formula C12H15NO2·C7H8O3S
Molecular Weight (g/mol) 377.45
MDL Number MFCD00044709
InChI Key NQLZTDKDXBKUGY-UHFFFAOYSA-N
Synonym 4-methylbenzene-1-sulfonic acid; 4-phenylpiperidine-4-carboxylic acid, 4-carboxy-4-phenylpiperidine tosylate, 4-phenylpiperidine-4-carboxylic acid compound with 4-methylbenzenesulfonic acid 1:1, 4-phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 4-phenyl-4-piperidinecarboxylic acid 4-methylbenzenesulfonate, 4-phenylpiperidine-4-carboxylic acid 4-toluenesulfonate, isonipecotic acid, 4-phenyl-; para-toluene sulfonate, 4-phenyl-4-carboxy piperidine tosylate, 4-carboxy-4-phenylpiperidine p-toluenesulfonate, 4-phenyl-piperidine-4-carboxylic acid tosylate
PubChem CID 2723657
IUPAC Name 4-methylbenzenesulfonic acid;4-phenylpiperidine-4-carboxylic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1CNCCC1(C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 225°C to 229°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information (3% in boiling water) Clear colorless to almost colorless
Formula Weight 377.45
Percent Purity 98%
Chemical Name or Material 4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%
compliance-icons
Product Identifier
  • 4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.