missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, MedChemExpress

Product Code. 16430758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16430758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16430758 Supplier Medchem Express Supplier No. HYA0281

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4-Phenylbutyric acid (4-PBA) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research.

4-Phenylbutyric acid (4-PBA) is an inhibitor of HDAC and endoplasmic reticulum (ER) stress, used in cancer and infection research.

Specifications

Chemical Name or Material 4-Phenylbutyric acid
Physical Form Powder
CAS 1821-12-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H12O2
For Use With (Application) Cancer-programmed cell death
Synonym 4-PBA Benzenebutyric acid
Solubility Information DMSO : ≥ 100 mg/mL (609.01 mM) ∣H2O : 2 mg/mL (12.18 mM; Need ultrasonic)
SMILES O=C(O)CCCC1=CC=CC=C1
Molecular Weight (g/mol) 164.2
Formula Weight 164.2
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.