missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcinnamic acid, 98%

Product Code. 11430493
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430493 50 g 50g
11420493 10 g 10g
11384679 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430493 Supplier Thermo Scientific Alfa Aesar Supplier No. A12643.18
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13026-23-8 | C15H12O2 | 224.26 g/mol

4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13026-23-8
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00014010
InChI Key DMJDEZUEYXVYNO-FLIBITNWSA-N
Synonym 4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid
PubChem CID 5842785
IUPAC Name (E)-3-(4-phenylphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 228°C
Quantity 50 g
Beilstein 1955124
Solubility Information Slightly soluble in water.
Formula Weight 224.27
Percent Purity 98%
Chemical Name or Material 4-Phenylcinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.