missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzaldehyde, 95%, Thermo Scientific™

Product Code. 10701684
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10701684 1 g Amber Glass Bottle 1g
10051924 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10701684 Supplier Thermo Scientific Maybridge Supplier No. CC56204DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 77232-38-3
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD08056282
InChI Key JUYLMPWKVWDXBE-UHFFFAOYSA-N
PubChem CID 12703722
SMILES O=CC1=CC=C(C=C1)C1=NC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl
IUPAC Name 4-pyrimidin-2-ylbenzaldehyde
Formula Weight 184.2
Percent Purity 95%
Chemical Name or Material 4-Pyrimidin-2-ylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.